Anti-CD366 (Tim-3) Rat Monoclonal Antibody [clone: B8.2C12]

Supplier: BioLegend
134002 134002
134002-BLEA 424.37 USD
CA134002-BL 134002-BL
Anti-CD366 (Tim-3) Rat Monoclonal Antibody [clone: B8.2C12]
Antibodies
Purified anti-mouse CD366 (Tim-3) [B8.2C12]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD366 (Tim-3)
Clonality: Monoclonal
Clone: B8.2C12
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR