Anti-NK-1.1 Mouse Monoclonal Antibody [clone: PK136]

Supplier: BioLegend
108702 108701 108702 108701
108702-BLEA 161.9 USD
108702-BL 108701-BL CA108702-BL CA108701-BL
Anti-NK-1.1 Mouse Monoclonal Antibody [clone: PK136]
Antibodies
Purified anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF®, IHC, IF, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: NK-1.1
Clonality: Monoclonal
Clone: PK136
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR