Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL2 Hamster Monoclonal Antibody () [clone: 4E2/MCP]

Supplier: Biolegend
506004
Purified anti-mouse MCP-1 [4E2/MCP]; Isotype: Hamster IgG; Reactivity: Mouse; Apps: ELISA Capture, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA capture antibody, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of MCP-1 recombinant protein ranging from 1000 to 8 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 4E2/MCP
Conjugation:
Epitope:
Host: Hamster
Isotype: Hamster IgG
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Conjugate Supplier No. VWR Catalog Number Unit Price Quantity
500 μg - 506004 506004-BL Each (500µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CCL2
Antigen Name CCL2
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 4E2/MCP
Host Hamster
Isotype Hamster IgG
Western Blot Yes
ELISA Yes
Gel Shift Yes
Antigen Synonyms Monocyte chemoattractant protein-1, MCAF, JE, SCYA2, HC-11, P6, SMC-CF, CCL2
Gene ID 20296
Concentration 0.50 mg/mL
Immunogen CHO-expressed, recombinant mouse MCP-1
Purification The antibody was purified by affinity chromatography.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty