Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8]

Supplier: BioLegend
127601 127601 127602 127602
127602-BLEA 169.39 USD
CA127601-BL 127602-BL CA127602-BL 127601-BL
Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8]
Antibodies
Purified anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G
Clonality: Monoclonal
Clone: 1A8
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR