Anti-JAML Armenian Hamster Monoclonal Antibody [clone: 4E10]

Supplier: BioLegend
128502 128502
128502-BLEA 443.52 USD
CA128502-BL 128502-BL
Anti-JAML Armenian Hamster Monoclonal Antibody [clone: 4E10]
Antibodies
Purified anti-mouse JAML [4E10]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: JAML
Clonality: Monoclonal
Clone: 4E10
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR