Anti-IL-21R Rat Monoclonal Antibody [clone: 4A9]

Supplier: BioLegend
131902 131902
131902-BLEA 380.01 USD
CA131902-BL 131902-BL
Anti-IL-21R Rat Monoclonal Antibody [clone: 4A9]
Antibodies
Purified anti-mouse IL-21 Receptor [4A9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-21R
Clonality: Monoclonal
Clone: 4A9
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR