Anti-IgG3 Rat Monoclonal Antibody [clone: RMG3-1]

Supplier: BioLegend
406802 406802
406802-BLEA 184.56 USD
CA406802-BL 406802-BL
Anti-IgG3 Rat Monoclonal Antibody [clone: RMG3-1]
Antibodies
Purified anti-mouse IgG3 [RMG3-1]; Isotype: Rat IgG; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For ELISA capture, the suggested use of this reagent is 2.0 μg per ml. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG3
Clonality: Monoclonal
Clone: RMG3-1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR