Anti-Ig light chain k Rat Monoclonal Antibody [clone: RMK-12]

Supplier: BioLegend
407202 407202
407202-BLEA 199.2 USD
CA407202-BL 407202-BL
Anti-Ig light chain k Rat Monoclonal Antibody [clone: RMK-12]
Antibodies
Purified anti-mouse Ig light chain κ [RMK-12]; Isotype: Rat IgG; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For ELISA capture, the suggested use of this reagent is 5.0 μg per ml. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ig light chain k
Clonality: Monoclonal
Clone: RMK-12
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR