Anti-IFN-b Armenian Hamster Monoclonal Antibody [clone: MIB-5E9.1]

Supplier: BioLegend
508102 508102
508102-BLEA 304.44 USD
508102-BL CA508102-BL
Anti-IFN-b Armenian Hamster Monoclonal Antibody [clone: MIB-5E9.1]
Antibodies
Purified anti-mouse IFN-β [MIB-5E9.1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: WB; Size: 500 μg

<p style="text-align: left;">Each lot of this antibody is quality control tested by <a title="View Protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_Western_blotting_protocol.pdf" target="_blank">Western blotting</a>. The suggested optimal usage is 1 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-b
Clonality: Monoclonal
Clone: MIB-5E9.1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR