Anti-I-Ab Mouse Monoclonal Antibody [clone: 25-9-17]

Supplier: BioLegend
114402 114402
114402-BLEA 273.08 USD
CA114402-BL 114402-BL
Anti-I-Ab Mouse Monoclonal Antibody [clone: 25-9-17]
Antibodies
Purified anti-mouse I-Ab (Aβb) [25-9-17]; Isotype: Mouse (C3H) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with alloantigens of H-2d,p,q haplotypes; Apps: FC, IHC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: I-Ab
Clonality: Monoclonal
Clone: 25-9-17
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse (C3H) IgG2a, κ
Reactivity: Mouse b haplotype;Cross-reacts with alloantigens of H-2d,p,q haplotypes
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR