Anti-H-2Kq Mouse Monoclonal Antibody [clone: KH114]

Supplier: BioLegend
115101 115101
115101-BLEA 89.58 USD
CA115101-BL 115101-BL
Anti-H-2Kq Mouse Monoclonal Antibody [clone: KH114]
Antibodies
Purified anti-mouse H-2Kq [KH114]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse q haplotype, Does not cross-react with other haplotypes (e.g. a, b, d, f, k, n, p, r, s, u, v); Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kq
Clonality: Monoclonal
Clone: KH114
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2a, κ
Reactivity: Mouse q haplotype;Does not cross-react with other haplotypes (e.g. a;b;d;f;k;n;p;r;s;u;v)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR