Anti-CDK5 Mouse Monoclonal Antibody [clone: 1321CT281.130.129]

Supplier: Abcepta
AM2261B
10218-100EA 593.03 USD
10218-100
Anti-CDK5 Mouse Monoclonal Antibody [clone: 1321CT281.130.129]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CDK5
Clonality: Monoclonal
Clone: 1321CT281.130.129
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR