Anti-CD54 Rat Monoclonal Antibody [clone: YN1/1.7.4]

Supplier: BioLegend
116102 116101 116101 116102
116102-BLEA 158.43 USD
CA116101-BL CA116102-BL 116102-BL 116101-BL
Anti-CD54 Rat Monoclonal Antibody [clone: YN1/1.7.4]
Antibodies
Purified anti-mouse CD54 [YN1/1.7.4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunohistochemical staining on frozen tissue sections, the suggested use is 5-10 μg/mL. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: YN1/1.7.4
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR