Anti-CD5 Rat Monoclonal Antibody [clone: 53-7.3]

Supplier: BioLegend
100602 100602
100602-BLEA 205.23 USD
CA100602-BL 100602-BL
Anti-CD5 Rat Monoclonal Antibody [clone: 53-7.3]
Antibodies
Purified anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF®, IHC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR