Anti-CD49e Rat Monoclonal Antibody [clone: 5H10-27(MFR5)]

Supplier: BioLegend
103801 103801
103801-BLEA 89.58 USD
103801-BL CA103801-BL
Anti-CD49e Rat Monoclonal Antibody [clone: 5H10-27(MFR5)]
Antibodies
Purified anti-mouse CD49e [5H10-27(MFR5)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49e
Clonality: Monoclonal
Clone: 5H10-27(MFR5)
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR