Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104]

Supplier: BioLegend
109802 109802
109802-BLEA 138.4 USD
109802-BL CA109802-BL
Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104]
Antibodies
Purified anti-mouse CD45.2 [104]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 104
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse (SJL) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR