Anti-CD34 Armenian Hamster Monoclonal Antibody [clone: HM34]

Supplier: BioLegend
128601 128601
128601-BLEA 76.64 USD
128601-BL CA128601-BL
Anti-CD34 Armenian Hamster Monoclonal Antibody [clone: HM34]
Antibodies
Purified anti-mouse CD34 [HM34]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: HM34
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR