Anti-P88945 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP15004A
89521-422EA 637.11 USD
89521-422
Anti-P88945 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: P88945
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR