Anti-CD200 R (OX2R) Rat Monoclonal Antibody [clone: OX-110]

Supplier: BioLegend
123902 123902
123902-BLEA 278.53 USD
CA123902-BL 123902-BL
Anti-CD200 R (OX2R) Rat Monoclonal Antibody [clone: OX-110]
Antibodies
Purified anti-mouse CD200R (OX2R) [OX-110]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD200 R (OX2R)
Clonality: Monoclonal
Clone: OX-110
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR