Anti-CD197 (CCR7) Rat Monoclonal Antibody [clone: 4B12]

Supplier: BioLegend
120101 120101
120101-BLEA 139.32 USD
120101-BL CA120101-BL
Anti-CD197 (CCR7) Rat Monoclonal Antibody [clone: 4B12]
Antibodies
Purified anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD197 (CCR7)
Clonality: Monoclonal
Clone: 4B12
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR