Anti-CD152 Syrian Hamster Monoclonal Antibody [clone: 9H10]

Supplier: BioLegend
106202 106202
106202-BLEA 152.22 USD
106202-BL CA106202-BL
Anti-CD152 Syrian Hamster Monoclonal Antibody [clone: 9H10]
Antibodies
Purified anti-mouse CD152 [9H10]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD152
Clonality: Monoclonal
Clone: 9H10
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: Syrian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR