Anti-MCM7 Rabbit Polyclonal Antibody [clone: Poly6027]

Supplier: BioLegend
602701 602702 602702 602701
602701-BLEA 88.57 USD
602701-BL 602702-BL CA602701-BL CA602702-BL
Anti-MCM7 Rabbit Polyclonal Antibody [clone: Poly6027]
Purified anti-MCM7 [Poly6027]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Human; Apps: WB; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MCM7
Clonality: Polyclonal
Clone: Poly6027
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR