Anti-MCM3 Rabbit Polyclonal Antibody [clone: Poly6023]

Supplier: BioLegend
602302 602302
602302-BLEA 258.15 USD
602302-BL CA602302-BL
Anti-MCM3 Rabbit Polyclonal Antibody [clone: Poly6023]
Purified anti-MCM3 [Poly6023]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Human; Apps: WB; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MCM3
Clonality: Polyclonal
Clone: Poly6023
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR