Anti-Lck Mouse Monoclonal Antibody [clone: LCK-01]

Supplier: BioLegend
628302 628301 628301 628302
628302-BLEA 247.16 USD
628302-BL 628301-BL CA628302-BL CA628301-BL
Anti-Lck Mouse Monoclonal Antibody [clone: LCK-01]
Antibodies
Purified anti-Lck [LCK-01]; Isotype: Mouse IgG1, κ; Reactivity: Human, does not cross-react with mouse; Apps: WB, IF, IP, FC, ICC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Lck
Clonality: Monoclonal
Clone: LCK-01
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;does not cross-react with mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR