Anti-LAT Mouse Monoclonal Antibody [clone: 11B.12]

Supplier: BioLegend
641102 641102
641102-BLEA 247.16 USD
641102-BL CA641102-BL
Anti-LAT Mouse Monoclonal Antibody [clone: 11B.12]
Antibodies
Purified anti-LAT [11B.12]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Human; Apps: WB, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: 11B.12
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR