Anti-KLH Mouse Monoclonal Antibody [clone: MTK2059]

Supplier: BioLegend
408402 408402 408401 408401
408401-BLEA 92.96 USD
CA408401-BL CA408402-BL 408401-BL 408402-BL
Anti-KLH Mouse Monoclonal Antibody [clone: MTK2059]
Antibodies
Purified anti-KLH [MTK2059]; Isotype: Mouse IgG1, κ; Reactivity: 2nd Step; Apps: ELISA; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For direct ELISA, the suggested use of this reagent is 0.5-2.0 μg/ml. To obtain a linear standard curve, serial dilutions of KLH ranging from 300 to 5 ng/ml are recommended for each ELISA. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: KLH
Clonality: Monoclonal
Clone: MTK2059
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: 2nd Step
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR