Anti-JNK1 Rabbit Polyclonal Antibody [clone: Poly6331]

Supplier: BioLegend
633101 633101 633102 633102
633102-BLEA 258.15 USD
633102-BL 633101-BL CA633102-BL CA633101-BL
Anti-JNK1 Rabbit Polyclonal Antibody [clone: Poly6331]
Purified anti-JNK1 [Poly6331]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, Mouse, Rat; Apps: WB, IP, IF; Size: 500 μl

Each lot of this antibody is quality control tested by Western blotting.  Western blotting suggested working dilution(s): Use 25 μl per 5 ml antibody dilution buffer for each mini-gel.It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone: Poly6331
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR