Anti-integrin b7 Rat Monoclonal Antibody [clone: FIB504]

Supplier: BioLegend
321202 321202
321202-BLEA 123.08 USD
321202-BL CA321202-BL
Anti-integrin b7 Rat Monoclonal Antibody [clone: FIB504]
Antibodies
Purified anti-human/mouse integrin β7 [FIB504]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: integrin b7
Clonality: Monoclonal
Clone: FIB504
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Mouse;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR