Anti-IL-21 Mouse Monoclonal Antibody [clone: 7H20-I19-M3]

Supplier: BioLegend
514302 514302
514302-BLEA 278.53 USD
CA514302-BL 514302-BL
Anti-IL-21 Mouse Monoclonal Antibody [clone: 7H20-I19-M3]
Antibodies
Purified anti-human/mouse IL-21 [7H20-I19-M3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 500 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-21
Clonality: Monoclonal
Clone: 7H20-I19-M3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR