Anti-TSLP Mouse Monoclonal Antibody [clone: 13H8.3]

Supplier: BioLegend
512102 512102
512102-BLEA 342.26 USD
CA512102-BL 512102-BL
Anti-TSLP Mouse Monoclonal Antibody [clone: 13H8.3]
Antibodies
Purified anti-human TSLP [13H8.3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.5-2 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of TSLP recombinant protein ranging from 200 to 3.1 ng/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TSLP
Clonality: Monoclonal
Clone: 13H8.3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR