Anti-MSCA-1 (MSC, W8B2) Mouse Monoclonal Antibody [clone: W8B2]

Supplier: BioLegend
327302 327302
327302-BLEA 222.3 USD
CA327302-BL 327302-BL
Anti-MSCA-1 (MSC, W8B2) Mouse Monoclonal Antibody [clone: W8B2]
Antibodies
Purified anti-human TNAP [W8B2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MSCA-1 (MSC, W8B2)
Clonality: Monoclonal
Clone: W8B2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR