Anti-IL-8 Mouse Monoclonal Antibody [clone: E8N1]

Supplier: BioLegend
511402 511402
511402-BLEA 304.44 USD
CA511402-BL 511402-BL
Anti-IL-8 Mouse Monoclonal Antibody [clone: E8N1]
Antibodies
Purified anti-human IL-8 [E8N1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.25-1.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-8 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-8
Clonality: Monoclonal
Clone: E8N1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR