Anti-VCP Mouse Monoclonal Antibody [clone: 1344CT150.163.114]

Supplier: Abcepta
AM2262B
10218-098EA 577.43 USD
10218-098
Anti-VCP Mouse Monoclonal Antibody [clone: 1344CT150.163.114]
Antibodies
Western Blot: 1:2000, Immunofluorescence: 0.0590277777777778,

Type: Primary
Antigen: VCP
Clonality: Monoclonal
Clone: 1344CT150.163.114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR