Anti-CD106 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 429 (MVCAM.A)]

Supplier: BioLegend
105714 105714 105713 105713
105714-BLEA 375.79 USD
105714-BL CA105714-BL 105713-BL CA105713-BL
Anti-CD106 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 429 (MVCAM.A)]
Antibodies
PE anti-mouse CD106 [429 (MVCAM.A)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD106
Clonality: Monoclonal
Clone: 429 (MVCAM.A)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR