Anti-GRN Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20450B
89513-480EA 637.11 USD
89513-480
Anti-GRN Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25,

Type: Primary
Antigen: GRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR