Anti-ARNT Rabbit Polyclonal Antibody

Supplier: Abcepta
AP14310C
10217-592EA 580.54 USD
10217-592
Anti-ARNT Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: ARNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR