PE anti-human IFN-γ R β chain [2HUB-159]; Isotype: Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.
Type: Primary
Antigen: IFN-g R b chain
Clonality: Monoclonal
Clone: 2HUB-159
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: Hamster IgG
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus