Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GITR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 621]

Supplier: Biolegend
311603 311604
PE anti-human CD357 (GITR) [621]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: GITR
Clonality: Monoclonal
Clone: 621
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 tests 311604 311604-BL Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 tests 311603 311603-BL Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GITR
Antigen Name GITR
Reactivity Cynomolgus, Human, Baboon, Rhesus Monkey, African Green Baboon, Human, Cynomolgus, African Green, Rhesus Monkey
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody Type Primary
Clone 621
Host Mouse
Isotype Mouse IgG1, κ
Flow Cytometry Yes
Antigen Synonyms Activation-inducible TNFR, AITR, Glucocorticoid-inducible TNFR, TNFRSF18
Gene ID 8784
Concentration 0.00 μg/mL
Purification The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. <B>Do not freeze.</b>
Shipping Temperature Ambient RT

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty