Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AER-37 (CRA-1)]

Supplier: BioLegend
334609 334610 334610 334609
334610-BLEA 368.02 USD
334610-BL CA334610-BL CA334609-BL 334609-BL
Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AER-37 (CRA-1)]
Antibodies
PE anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <b>Test size products are transitioning from 20 μl to 5 μl per test</b>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: Fc epsilon RI alpha
Clonality: Monoclonal
Clone: AER-37 (CRA-1)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR