Anti-NFYB Rabbit Polyclonal Antibody

Supplier: Abcepta
AP14334A
89512-372EA 636.79 USD
89512-372
Anti-NFYB Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: NFYB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR