Anti-EIF4B Mouse Monoclonal Antibody [clone: 355CT12.6.5]

Supplier: Abcepta
AM1962A
89521-236EA 606.77 USD
89521-236
Anti-EIF4B Mouse Monoclonal Antibody [clone: 355CT12.6.5]
Antibodies
Western Blot: 1:500~4000,

Type: Primary
Antigen: EIF4B
Clonality: Monoclonal
Clone: 355CT12.6.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR