Anti-AHCY Mouse Monoclonal Antibody [clone: 445CT13.3.3]

Supplier: Abcepta
AM2030A
89512-194EA 637.11 USD
89512-194
Anti-AHCY Mouse Monoclonal Antibody [clone: 445CT13.3.3]
Antibodies
Western Blot: 1:1000~8000,

Type: Primary
Antigen: AHCY
Clonality: Monoclonal
Clone: 445CT13.3.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR