Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OKT4]

Supplier: BioLegend
317409 317409 317410 317410
317410-BLEA 90.04 USD
CA317409-BL CA317410-BL 317410-BL 317409-BL
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OKT4]
Antibodies
PE anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR