Anti-CD284 (TLR4) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HTA125]

Supplier: BioLegend
312806 312805 312805 312806
312805-BLEA 131.66 USD
CA312805-BL 312805-BL 312806-BL CA312806-BL
Anti-CD284 (TLR4) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HTA125]
Antibodies
PE anti-human CD284 (TLR4) [HTA125]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Guinea Pig; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: CD284 (TLR4)
Clonality: Monoclonal
Clone: HTA125
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR