Anti-CD33 Mouse Monoclonal Antibody [clone: 574CT16.4.4]

Supplier: Abcepta
AM2149B
89514-968EA 637.11 USD
89514-968
Anti-CD33 Mouse Monoclonal Antibody [clone: 574CT16.4.4]
Antibodies
Western Blot: 1:100~500, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: 574CT16.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR