Anti-CD48 Armenian Hamster Monoclonal Antibody (LEAF) [clone: HM48-1]

103408 103408
103408-BLEA 290.47 USD
103408-BL CA103408-BL
Anti-CD48 Armenian Hamster Monoclonal Antibody (LEAF) [clone: HM48-1]
LEAF™ Purified anti-mouse CD48 [HM48-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IP, Costim, Block; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: HM48-1
Conjugation: LEAF
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR