Anti-CD19 Rat Monoclonal Antibody (LEAF) [clone: 6D5]

115514 115514
115514-BLEA 286.06 USD
115514-BL CA115514-BL
Anti-CD19 Rat Monoclonal Antibody (LEAF) [clone: 6D5]
LEAF™ Purified anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF®, IF; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: LEAF
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR