Anti-CD49d Rat Monoclonal Antibody (Biotin) [clone: 9C10 (MFR4.B)]

Supplier: BioLegend
103703 103703
103703-BLEA 96.04 USD
CA103703-BL 103703-BL
Anti-CD49d Rat Monoclonal Antibody (Biotin) [clone: 9C10 (MFR4.B)]
Antibodies
Biotin anti-mouse CD49d [9C10 (MFR4.B)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: 9C10 (MFR4.B)
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR