Anti-CD4 Rat Monoclonal Antibody (Biotin) [clone: RM4-5]

Supplier: BioLegend
100507 100507 100508 100508
100507-BLEA 38.86 USD
CA100508-BL CA100507-BL 100507-BL 100508-BL
Anti-CD4 Rat Monoclonal Antibody (Biotin) [clone: RM4-5]
Antibodies
Biotin anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR