Anti-CD36 Armenian Hamster Monoclonal Antibody (Biotin) [clone: HM36]

Supplier: BioLegend
102604 102604
102604-BLEA 348.72 USD
102604-BL CA102604-BL
Anti-CD36 Armenian Hamster Monoclonal Antibody (Biotin) [clone: HM36]
Antibodies
Biotin anti-mouse CD36 [HM36]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: HM36
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR